Learn More
Eisco™ Hooked Iron Weight
Hooked cast iron hooked weight features hook for easy lifting and hanging and bottom slot to allow other hooked weights to be attached in tandem
$12.89 - $34.21
Specifications
Product Type Hooked Weight
Material Cast Iron
Includes Bottom Slot
View More Specs
Products 6
Catalog Number Mfr. No. Weight (Metric) Price Quantity & Availability  
Catalog Number Mfr. No. Weight (Metric) Price Quantity & Availability  
S27628
View Documents Promotion Details
Eisco™
HKW10G
10 g
Each for $14.46
Only null left
S27630
View Documents Promotion Details
Eisco™
HKW20G
20 g
Each for $14.46
Only null left
S27632
View Documents Promotion Details
Eisco™
HKW50G
50 g
Each for $12.89
Only null left
S27627
View Documents Promotion Details
Eisco™
HKW100
100 g
Each for $17.57
Only null left
S27629
View Documents Promotion Details
Eisco™
HKW200
200 g
Each for $23.07
Only null left
S27631
View Documents Promotion Details
Eisco™
HKW500
500 g
Each for $34.21
Only null left
Description
Description
  • Single hooked weight
  • Made from high quality cast iron
  • Cylindrical with cone shaped top
  • Hook for easy lifting and hanging
  • Bottom slot to allow other hooked weights to be attached in tandem
Specifications
Specifications
Hooked Weight
Cast Iron
Bottom Slot