Learn More
Eisco™ Hooked Iron Weight
Description
- Single hooked weight
- Made from high quality cast iron
- Cylindrical with cone shaped top
- Hook for easy lifting and hanging
- Bottom slot to allow other hooked weights to be attached in tandem
Specifications
Specifications
| Product Type | Hooked Weight |
| Material | Cast Iron |
| Weight (Metric) | 20 g |
| Includes | Bottom Slot |