Learn More
Eisco™ Hooked Iron Weight
Catalog No. S27632
Change view
Click to view available options
Weight (Metric):
10 g
20 g
50 g
100 g
200 g
500 g
6 product options available for selection
Product selection table with 6 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Weight (Metric)
S27632 50 g
S27628 10 g
S27630 20 g
S27627 100 g
S27629 200 g
S27631 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 6 options available.
6 options
Catalog No. S27632 Supplier Eisco™ Supplier No. HKW50G
Hooked cast iron hooked weight features hook for easy lifting and hanging and bottom slot to allow other hooked weights to be attached in tandem
  • Single hooked weight
  • Made from high quality cast iron
  • Cylindrical with cone shaped top
  • Hook for easy lifting and hanging
  • Bottom slot to allow other hooked weights to be attached in tandem
Specifications
Product Type Hooked Weight
Material Cast Iron
Weight (Metric) 50 g
Includes Bottom Slot