missing translation for 'onlineSavingsMsg'
Learn More

Honokiol, 98+%

Catalog No. p-7042704
Change view
Click to view available options
Quantity:
10 mg
25 mg
100 mg
Catalog No. Quantity
AAJ63434MA 10 mg
AAJ63434MB 25 mg
AAJ63434MC 100 mg
3 options

Catalog No. AAJ63434MA

Supplier: Thermo Scientific Chemicals J63434MA

Only null left

Induces apoptosis in human squamous lung cancer cells | CAS: 35354-74-6 | C18H18O2 | 266.34 g/mol

Induces apoptosis in human squamous lung cancer cells

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 35354-74-6
Molecular Formula C18H18O2
Molecular Weight (g/mol) 266.34
MDL Number MFCD00016674
InChI Key FVYXIJYOAGAUQK-UHFFFAOYSA-N
Synonym honokiol, 5,3'-diallyl-2,4'-dihydroxybiphenyl, 3,5'-diallyl-4,2'-dihydroxybiphenyl, unii-11513cco0n, chembl16901, 3',5-diallyl-1,1'-biphenyl-2,4'-diol, 4-allyl-2-3-allyl-4-hydroxy-phenyl phenol, 3',5-diallyl-2,4'-biphenyldiol, 3',5-diallylbiphenyl-2,4'-diol, 5,3′-diallyl-2,4′-dihydroxydiphenyl
PubChem CID 72303
ChEBI CHEBI:5759
IUPAC Name 2-(4-hydroxy-3-prop-2-enylphenyl)-4-prop-2-enylphenol
SMILES C=CCC1=CC(=C(C=C1)O)C2=CC(=C(C=C2)O)CC=C

Specifications

Melting Point 86°C
Color White
Quantity 10 mg
UN Number UN3077
Merck Index 14,4742
Formula Weight 266.34
Percent Purity ≥98%
Physical Form Powder
Chemical Name or Material Honokiol
Hazard Category H318
Hazard Statement GHS H Statement
H318
Causes serious eye damage.
Precautionary Statement P280-P305+P351+P338-P310
DOTInformation Transport Hazard Class: 9; Packing Group: III; Proper Shipping Name: ENVIRONMENTALLY HAZARDOUS SUBSTANCES, SOLID, N.O.S.
TSCA No
Recommended Storage Keep cold

RUO – Research Use Only