missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Homogentisic Acid gamma-Lactone 98.0+%, TCI America™
$177.08 - $488.38
Chemical Identifiers
| CAS | 2688-48-4 |
|---|---|
| Molecular Formula | C8H6O3 |
| Molecular Weight (g/mol) | 150.133 |
| MDL Number | MFCD00005857 |
| InChI Key | POUITAHNNRJWMA-UHFFFAOYSA-N |
| Synonym | 5-hydroxybenzofuran-2 3h-one, homogentisic lactone, homogentisic acid gamma-lactone, 5-hydroxybenzofuran-2-one, 5-hydroxybenzofuran-2-one, 5-hydroxy-2 3h-benzofuranone, 5-hydroxy-2 3h-benzofuranone, homogentisic acid, homogentisic acid, 5-hydroxy-3-hydrobenzo b furan-2-one, 5-hydroxy-3-hydrobenzo b furan-2-one |
| PubChem CID | 75898 |
| IUPAC Name | 5-hydroxy-3H-1-benzofuran-2-one |
| SMILES | C1C2=C(C=CC(=C2)O)OC1=O |
Chemical Identifiers
| 2688-48-4 | |
| 150.133 | |
| POUITAHNNRJWMA-UHFFFAOYSA-N | |
| 75898 | |
| C1C2=C(C=CC(=C2)O)OC1=O |
| C8H6O3 | |
| MFCD00005857 | |
| 5-hydroxybenzofuran-2 3h-one, homogentisic lactone, homogentisic acid gamma-lactone, 5-hydroxybenzofuran-2-one, 5-hydroxybenzofuran-2-one, 5-hydroxy-2 3h-benzofuranone, 5-hydroxy-2 3h-benzofuranone, homogentisic acid, homogentisic acid, 5-hydroxy-3-hydrobenzo b furan-2-one, 5-hydroxy-3-hydrobenzo b furan-2-one | |
| 5-hydroxy-3H-1-benzofuran-2-one |
Specifications
| 2688-48-4 | |
| White-Yellow | |
| MFCD00005857 | |
| 5-hydroxybenzofuran-2 3h-one, homogentisic lactone, homogentisic acid gamma-lactone, 5-hydroxybenzofuran-2-one, 5-hydroxybenzofuran-2-one, 5-hydroxy-2 3h-benzofuranone, 5-hydroxy-2 3h-benzofuranone, homogentisic acid, homogentisic acid, 5-hydroxy-3-hydrobenzo b furan-2-one, 5-hydroxy-3-hydrobenzo b furan-2-one | |
| C1C2=C(C=CC(=C2)O)OC1=O | |
| 150.133 | |
| 150.13 | |
| Crystalline Powder |
| 193°C | |
| C8H6O3 | |
| 1 g | |
| POUITAHNNRJWMA-UHFFFAOYSA-N | |
| 5-hydroxy-3H-1-benzofuran-2-one | |
| 75898 | |
| ≥98.0% (GC) | |
| Homogentisic Acid gamma-Lactone |
Safety and Handling
TSCA : No