CAS RN 2688-48-4
IUPAC Name:
5-hydroxy-2,3-dihydro-1-benzofuran-2-one
Synonyms:
5-hydroxy-3H-1-benzofuran-2-one
Molecular Weight (g/mol):
150.13
Molecular Formula:
C8H6O3
InChi Key:
POUITAHNNRJWMA-UHFFFAOYSA-N
SMILES:
OC1=CC=C2OC(=O)CC2=C1
1
–
1
of
1
results
1
Homogentisic Acid gamma-Lactone 98.0+%, TCI America™
CAS: 2688-48-4 Molecular Formula: C8H6O3 Molecular Weight (g/mol): 150.133 MDL Number: MFCD00005857 InChI Key: POUITAHNNRJWMA-UHFFFAOYSA-N Synonym: 5-hydroxybenzofuran-2 3h-one,5-hydroxy-2-coumaranone,homogentisic acid gamma-lactone,5-hydroxybenzofuran-2-one,2,3-dihydro-5-hydroxybenzofuran-2-one,5-hydroxy-2 3h-benzofuranone,2 3h-benzofuranone, 5-hydroxy,homogentisic acid,a-lactone,5-hydroxy-3-hydrobenzo b furan-2-one,homogentisic lactone PubChem CID: 75898 IUPAC Name: 5-hydroxy-3H-1-benzofuran-2-one SMILES: C1C2=C(C=CC(=C2)O)OC1=O