missing translation for 'onlineSavingsMsg'
Learn More

Homogentisic Acid gamma-Lactone 98.0+%, TCI America™

Catalog No. H06305G
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
H06305G 5 g
H06301G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. H06305G Supplier TCI America Supplier No. H06305G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 2688-48-4
Molecular Formula C8H6O3
Molecular Weight (g/mol) 150.133
MDL Number MFCD00005857
InChI Key POUITAHNNRJWMA-UHFFFAOYSA-N
Synonym 5-hydroxybenzofuran-2 3h-one, 5-hydroxy-2-coumaranone, homogentisic acid gamma-lactone, 5-hydroxybenzofuran-2-one, 2,3-dihydro-5-hydroxybenzofuran-2-one, 5-hydroxy-2 3h-benzofuranone, 2 3h-benzofuranone, 5-hydroxy, homogentisic acid, a-lactone, 5-hydroxy-3-hydrobenzo b furan-2-one, homogentisic lactone
PubChem CID 75898
IUPAC Name 5-hydroxy-3H-1-benzofuran-2-one
SMILES C1C2=C(C=CC(=C2)O)OC1=O

Specifications

Melting Point 193°C
Color White-Yellow
Quantity 5 g
Formula Weight 150.13
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material Homogentisic Acid gamma-Lactone