missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Holmium rod, 6.35mm (0.25in) dia, 99.8% (metals basis)
CAS: 7440-60-0 | Ho | 164.93 g/mol
Supplier: Thermo Scientific Chemicals 44050DJ
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
- Packaged in mylar under argon
Specifications
| Holmium rod | |
| 7440-60-0 | |
| Odorless | |
| ≈0.28g/mm | |
| (metals basis) | |
| MFCD00011049 | |
| KJZYNXUDTRRSPN-UHFFFAOYSA-N | |
| holmium | |
| 23988 | |
| 99.9% |
| ≈6.35mm (0.25 in.) dia. | |
| 50 mm | |
| Mylar | |
| Cast Rod | |
| Ho | |
| atom, unii-w1xx32sqn1, w1xx32sqn1, foil, holmio, chips, ingot, pieces, powder, acmc-1c9ym | |
| [Ho] | |
| 164.93 | |
| CHEBI:49648 |
Chemical Identifiers
| 7440-60-0 | |
| 164.93 | |
| KJZYNXUDTRRSPN-UHFFFAOYSA-N | |
| 23988 | |
| holmium |
| Ho | |
| MFCD00011049 | |
| atom, unii-w1xx32sqn1, w1xx32sqn1, foil, holmio, chips, ingot, pieces, powder, acmc-1c9ym | |
| CHEBI:49648 | |
| [Ho] |
Safety and Handling
EINECSNumber : 231-169-0
TSCA : Yes
storageNote1 :
- Packaged in mylar under argon
Recommended Storage : Ambient temperatures
RUO – Research Use Only