missing translation for 'onlineSavingsMsg'
Learn More

Holmium rod, 6.35mm (0.25in) dia, 99.8% (metals basis)

Catalog No. p-7059139
Change view
Click to view available options
Quantity:
10 mm
50 mm
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AA44050HF 10 mm
AA44050DJ 50 mm
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AA44050HF Supplier Thermo Scientific Chemicals Supplier No. 044050HF
Only null left

CAS: 7440-60-0 | Ho | 164.93 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

  • Packaged in mylar under argon

Chemical Identifiers

CAS 7440-60-0
Molecular Formula Ho
Molecular Weight (g/mol) 164.93
MDL Number MFCD00011049
InChI Key KJZYNXUDTRRSPN-UHFFFAOYSA-N
Synonym atom, unii-w1xx32sqn1, w1xx32sqn1, foil, holmio, chips, ingot, pieces, powder, acmc-1c9ym
PubChem CID 23988
ChEBI CHEBI:49648
IUPAC Name holmium
SMILES [Ho]

Specifications

Quantity 10 mm
Odor Odorless
Packaging Mylar under Argon
Weight ≈0.28g/mm
Form Cast Rod
Percent Purity 99.9%
Assay (metals basis)
Chemical Name or Material Holmium rod, ≈6.35mm (0.25 in.) dia.
EINECSNumber 231-169-0
TSCA Yes
storageNote1
  • Packaged in mylar under argon
Recommended Storage Ambient temperatures

RUO – Research Use Only