missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Hoefer™ Glass Plates for Vertical Electrophoresis Units
Glass plates for vertical electrophoresis units
Supplier: Hoefer™ SE6602
Description
Glass plates for vertical electrophoresis units
- For use with Hoefer SE600 and SE400 Series Vertical Electrophoresis Units
- Storage temperature: +15°C to +30°C
Specifications
| SE 600, SE 400 Series Vertical Electrophoresis System | |
| Glass | |
| 9.4 in. |
| Glass Plate | |
| 18 x 24cm |