missing translation for 'onlineSavingsMsg'
Learn More

Hoefer™ Glass Plates for Vertical Electrophoresis Units DFS Item

Glass plates for vertical electrophoresis units

$249.24 - $340.44

Specifications

For Use With (Equipment) SE 600, SE 400 Series Vertical Electrophoresis System
Type Glass Plate
Material Glass
View More Specs

Products 2
Catalog Number Mfr. No. Dimensions (L x W) Price Quantity  
Catalog Number Mfr. No. Dimensions (L x W) Price Quantity  
36-100-3837 DFS Item
View Documents
Hoefer™
SE6402
18 x 8cm
Pack of 2 Each for $249.24
Only null left
 
SE6602 DFS Item
View Documents
Hoefer™
SE6602
18 x 24cm
Each for $340.44
Only null left
 
Description

Description

Glass plates for vertical electrophoresis units

  • For use with Hoefer SE600 and SE400 Series Vertical Electrophoresis Units
  • Storage temperature: +15°C to +30°C
Specifications

Specifications

SE 600, SE 400 Series Vertical Electrophoresis System
Glass Plate
Glass
Videos
SDS
Documents

Documents