Learn More
Hoefer™ Combs for SE 600 Series or SE 400 Series Vertical Electrophoresis Units
Made from plastics that are highly resistant to acids, bases, and all commonly used buffer systems
$113.91 - $284.78
Specifications
| For Use With (Equipment) | SE 600, SE 400 Series Vertical Electrophoresis System |
|---|---|
| Material | PTFE |
| Depth (English) | 0.98 in. |
| Depth (Metric) | 2.5 cm |
| Catalog Number | Mfr. No. | Type | No. of Wells | Thickness (Metric) | Volume (Metric) Well | Price | Quantity | ||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Type | No. of Wells | Thickness (Metric) | Volume (Metric) Well | Price | Quantity | ||
03-500-237
|
Hoefer™
SE5111210 |
Comb | 12 | 0.1 cm | 7.7μL/ mm |
Each for $248.81
|
|
||
03-500-200
|
Hoefer™
SE511DR15 |
Comb | 1 Prep, 2 Reference | 0.15 cm | 171μL/ mm |
Each for $281.78
|
|
||
36-100-3852
|
Hoefer™
SE511DR.75 |
Comb | 1 Prep, 2 Reference | 0.07 cm | 85μL/ mm |
Each for $284.78
|
|
||
03-500-191
|
Hoefer™
SE5112075 |
Standard Comb | 20 | 0.07 cm | 3.1μL/ mm |
Each for $248.81
|
|
||
03-500-190
|
Hoefer™
SE511151.5 |
Standard Comb | 15 | 0.15 cm | 8.6μL/ mm |
Each for $248.81
|
|
||
03-500-193
|
Hoefer™
SE5112015 |
Standard Comb | 20 | 0.15 cm | 6.2μL/ mm |
Each for $248.81
|
|
||
03-500-188
|
Hoefer™
SE5111575 |
Standard Comb | 15 | 0.07 cm | 4.3μL/ mm |
Each for $248.81
|
|
||
36-100-3867
|
Hoefer™
SE51110.75 |
Standard Comb | 10 | 0.07 cm | 6.2μL/ mm |
Each for $248.81
|
|
||
03-500-187
|
Hoefer™
SE5111215 |
Standard Comb | 12 | 0.15 cm | 11.5μL/ mm |
Each for $248.81
|
|
||
03-500-186
|
Hoefer™
SE5111275 |
Standard Comb | 12 | 0.07 cm | 5.8μL/ mm |
Each for $248.81
|
|
||
36-100-3865
|
Hoefer™
SE511101.5 |
Standard Comb | 10 | 0.15 cm | 12.4μL/ mm |
Each for $248.81
|
|
||
03-500-197
|
Hoefer™
SE511BKA |
Adjustable Comb Back |
Each for $113.91
|
|
|||||
36-100-3849
|
Hoefer™
SE511R75 |
Comb | 1 Prep, 1 Reference | 0.07 cm | 90μL/ mm |
Each for $281.78
|
|
||
03-500-203
|
Hoefer™
SE511R15 |
Comb | 1 Prep, 1 Reference | 0.15 cm | 183μL/ mm |
Each for $281.78
|
|
||
03-500-202
|
Hoefer™
SE511R10 |
Comb | 1 Prep, 1 Reference | 0.1 cm | 120μL/ mm |
Each for $281.78
|
|
||
36-100-3856
|
Hoefer™
SE51128.75 |
Comb | 28 | 0.07 cm | 2.1μL/ mm |
Each for $248.81
|
|
||
03-500-196
|
Hoefer™
SE5112815 |
Comb | 28 | 0.15 cm | 4.1μL/ mm |
Each for $248.81
|
|
||
03-500-192
|
Hoefer™
SE5112010 |
Comb | 20 | 0.1 cm | 4.1μL/ mm |
Each for $248.81
|
|
||
03-500-189
|
Hoefer™
SE511-15-1.0 |
Comb | 15 | 0.1 cm | 5.7μL/ mm |
Each for $248.81
|
|
||
36-100-3866
|
Hoefer™
SE511101.0 |
Comb | 10 | 0.1 cm | 8.3μL/ mm |
Each for $248.81
|
|
||
Description
- Lightweight
- Virtually unbreakable
- Come attached to a standard back which produces maximum well depths of 25 mm, except for the 28-well preparative combs which produce well depths of 15 mm
- Adjustable comb back (available separately, below) converts 25 mm deep combs to 10 or 15 mm depth
Specifications
| SE 600, SE 400 Series Vertical Electrophoresis System | |
| PTFE | |
| 0.98 in. | |
| 2.5 cm |