Learn More
Hoefer™ Combs for SE 600 Series or SE 400 Series Vertical Electrophoresis Units
Made from plastics that are highly resistant to acids, bases, and all commonly used buffer systems
Supplier: Hoefer™ SE511R15
Description
- Lightweight
- Virtually unbreakable
- Come attached to a standard back which produces maximum well depths of 25 mm, except for the 28-well preparative combs which produce well depths of 15 mm
- Adjustable comb back (available separately, below) converts 25 mm deep combs to 10 or 15 mm depth
Specifications
| SE 600, SE 400 Series Vertical Electrophoresis System | |
| PTFE | |
| 0.05 in. | |
| 183μL/ mm | |
| 12.1 cm | |
| 2.5 cm |
| Comb | |
| 1 Prep, 1 Reference | |
| 0.15 cm | |
| 4.7 in. | |
| 0.98 in. |