missing translation for 'onlineSavingsMsg'
Learn More

Cytiva HiTrap™ Protein L Prepacked Columns

Catalog No. 45002605
Change view
Click to view available options
Quantity:
1 x 1mL
1 x 5mL
5 x 1mL
Diameter (English) Inner:
0.27 in.
0.62 in.
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Diameter (English) Inner
45002605 1 x 5mL 0.62 in.
45002606 5 x 1mL 0.27 in.
45000510 1 x 1mL 0.27 in.
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. 45002605 Supplier Cytiva Supplier No. 17547815

For capture of antibodies and antibody fragments such as Fab fragments, scFv, and Dabs

  • Prepacked with Capto L affinity chromatography medium (resin)
  • Polypropylene column are easily connected in series for increased productivity
  • Easy screening and convenient process development
  • For use with syringe, peristaltic pump, or chromatography system
  • Maximum pressure: 5 bar
  • 1 and 5mL columns

BioProcess Media
  • Ligand: Recombinant protein L (E. coli), mammalian free
  • Matrix: Rigid, highly cross-linked agarose
  • Average particle size: 85μm
  • pH stability working range: 2 to 10

1mL columns

  • Column I.D.: 7mm
  • Flow velocity: <500cm/h3

5mL columns
  • Column I.D.: 16mm
  • Flow velocity: <500cm/h4

Specifications

Product Type HiTrap Protein L column
Bed Volume 5mL
Diameter (English) Inner 0.62 in.
Diameter (Metric) Inner 16mm
Quantity 1 x 5mL
Height (English) 0.78 in.
Height (Metric) 20mm
Flow Rate <500 cm/hr.
Material Polypropylene
Ligand Type Recombinant Protein L (E. coli), mammalian free
Max. Pressure 5 bar
pH Range 2 to 10
Particle Size 85μm
Support Type Highly Cross-linked Agarose
Content And Storage 2° to 8°C
Column Type Affinity Column
For Use With (Application) Affinity chromato graphy medium (Resin) for capture of Antibodies and Antibody fragments such as Fab fragments, scFv and Dabs
Show More Show Less