missing translation for 'onlineSavingsMsg'
Learn More

Cytiva HiTrap™ MabSelect™ SuRe Prepacked Columns

Use these columns in small-scale MAb purification in a continuous bioprocess. Cytiva Life Sciences™ HiTrap™ MabSelect™ SuRe (Superior Resistance) Prepacked Columns for fast purification of monoclonal antibodies (Mabs) from large sample volumes

$576.81 - $6780.57

Specifications

Binding Capacity ≈35mg Human lgG/mL at 2.4 min. residence time
Content And Storage 4°C to 30°C, 20% Ethanol
For Use With (Application) Chromatography System, Manual
Type Pre-packed Column
View More Specs

Products 4
Catalog Number Mfr. No. Bed Volume Flow Rate Price Quantity  
Catalog Number Mfr. No. Bed Volume Flow Rate Price Quantity  
45000010
View Documents
Cytiva
11003495
5 mL <20 mL/min.
Pack of 5 Each for $6,780.57
Only null left
 
45-000-008
View Documents
Cytiva
11003493
1 mL <4 mL/min.
Pack of 5 Each for $1,736.35
Only null left
 
45-000-009
View Documents
Cytiva
11003494
5 mL <20 mL/min.
Each for $1,620.89
Only null left
 
45-000-512
View Documents
Cytiva
29049104
1 mL <4 mL/min.
Each for $576.81
Only null left
 
Description

Description

Prepacked HiTrap columns for process development, screening of purification conditions, and small-scale purification of Mabs. MabSelect media are suited for the high flowrates and pressures required when scaling up.

  • Oriented coupling of recombinant Protein A ligand to the agarose matrix via an engineered C-terminal cysteine increases IgG binding capacity over other protein A media
  • High capacity, low ligand leakage, and high-flow base matrix allow purification of Mabs at process scale
  • Fewer regulatory concerns due to the total absence of mammalian culture in the ligand production and purification
  • Designed for use with a syringe, a peristaltic pump, or a chromatography system

MabSelect

  • Allows at least five times higher flowrates than conventional cross-linked agarose of similar porosity
  • Recombinant Protein A ligand promotes specificity of binding to Fc region of IgG; permits one-step purification
MabSelect Xtra™
  • Same recombinant protein A ligand as MabSelect, but with a smaller particle size and greater porosity, for increased dynamic binding capacity
  • Allows binding from samples with high expression levels of the monoclonal antibody
  • Lowers overall production cost due to the possibility of processing concentrated feedstocks in fewer batches
MabSelect SuRe™ (Superior Resistance)
  • Akali-tolerant protein A-derived ligand allows the use of up to 0.5M NaOH for cleaning-in-place (CIP)
  • Permits significant cost savings and extension of cycles in regular production

Specifications

Specifications

≈35mg Human lgG/mL at 2.4 min. residence time
4°C to 30°C, 20% Ethanol
Chromatography System, Manual
Pre-packed Column
Videos
SDS
missing translation for 'documents'

missing translation for 'documents'