Learn More
Cytiva HiTrap MabSelect PrismA Column
Protein A Column
Supplier: Cytiva 17549852
Description
These columns are prepacked with MabSelect PrismA protein A chromatography resin that has been improved with an optimized high-flow agarose base matrix and a genetically engineered protein A-derived ligand, allowing efficient cleaning between purification runs.
Specifications
| 1 mL | |
| 5 bar (0.5 MPa, 70 psi) | |
| Antibody affinity chromatography |
| <4 mL/min. | |
| 5 x 1 mL | |
| Pre-packed Column |