missing translation for 'onlineSavingsMsg'
Learn More

Cytiva HiTrap MabSelect PrismA Column

Catalog No. 09928068
Change view
Click to view available options
Quantity:
1 x 1 mL
1 x 5 mL
5 x 1 mL
5 x 5 mL
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
09928068 1 x 1 mL
09928069 5 x 1 mL
09928070 1 x 5 mL
09928071 5 x 5 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. 09928068 Supplier Cytiva Supplier No. 17549851

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Protein A Column

These columns are prepacked with MabSelect PrismA protein A chromatography resin that has been improved with an optimized high-flow agarose base matrix and a genetically engineered protein A-derived ligand, allowing efficient cleaning between purification runs.

Specifications

Bed Volume 1 mL
Flow Rate <4 mL/min.
Max. Operating Pressure 5 bar (0.5 MPa, 70 psi)
Quantity 1 x 1 mL
For Use With (Application) Antibody affinity chromatography
Type Pre-packed Column