missing translation for 'onlineSavingsMsg'
Learn More

Cytiva™ HiTrap™ Chromatography Columns with MabSelect™ VL Protein L Resin

Catalog No. 01183310
Change view
Click to view available options
Quantity:
1 x 1 mL
1 x 5 mL
5 x 1 mL
5 x 5 mL
Bed Volume:
1 mL
5 mL
Diameter (Metric) Inner:
7 mm
16 mm
Flow Rate:
<20 mL/min.
<4 mL/min.
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Bed Volume Diameter (Metric) Inner Flow Rate
01183310 5 x 5 mL 5 mL 16 mm <20 mL/min.
01183307 1 x 1 mL 1 mL 7 mm <4 mL/min.
01183308 5 x 1 mL 1 mL 7 mm <4 mL/min.
01183309 1 x 5 mL 5 mL 16 mm <20 mL/min.
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. 01183310 Supplier Cytiva™ Supplier No. 17542054
Only null left

Protein L affinity chromatography columns for capture of antibody Fabs, bispecifics, and other variants containing a kappa light chain.

  • Redesigned protein L ligand captures antibody Fabs, bispecifics, and other variants containing a kappa light chain with high binding capacity and improved alkaline stability
  • High dynamic binding capacity for efficient capture of target molecules
  • Stable when cleaned with 0.1 M NaOH, reducing risk for bioburden incidents
  • Provides good resolution for product-related impurities in the capture of bispecific antibodies

As the diversity of antibody variants increases, so does the need for chromatography resins that can capture and purify these variants. As an alternative to protein A, protein L can be used to capture antibody Fabs, bispecific antibodies, and other molecules containing kappa variants of the variable light (VL) chain. Protein L binds human kappa light chain subtypes 1, 3, and 4 and mouse kappa light chain subtype 1.

Specifications

Bed Volume 5 mL
Diameter (English) Inner 0.63 in.
Diameter (Metric) Inner 16 mm
Quantity 5 x 5 mL
Height (English) 1 in.
Height (Metric) 25 mm
Flow Rate <20 mL/min.
Max. Pressure 5 bar
Column Type Affinity Column
For Use With (Application) Antibody Affinity Chromatography