missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Cytiva HiTrap Capto S ImpAct Prepacked Columns
HiTrap Capto S ImpAct columns are prepacked with Capto S ImpAct cation exchange chromatography medium for screening and small-scale protein purifications using ion exchange chromatography (IEX).
$411.02 - $1850.32
Specifications
| Product Type | HiTrap Capto S ImpAct Prepacked Column |
|---|---|
| Material | High-flow agarose |
| pH Range | 4 to 12 |
| Particle Size | 50μm |
| Column Type | Ion Exchange Column |
| Catalog Number | Mfr. No. | Quantity | Flow Rate | Price | Quantity | ||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Flow Rate | Price | Quantity | ||||
|
45-003-202
|
Cytiva
17371751 |
5 x 1mL | 4 mL/min. |
Each for $411.02
|
|
||||
|
45-003-203
|
Cytiva
17371755 |
5 x 5mL | 20 mL/min. |
Each for $1,850.32
|
|
||||
Description
- Convenient prepacked column for fast, reproducible, and easy small-scale purifications.
- High binding capacity, > 100 mg MAb/mL medium.
- Efficient aggregate removal at high load of monoclonal antibodies.
- High-resolution intermediate purification and polishing based on the well-established Capto platform.
- Flexibility of design thanks to a large operational window of flow rates and bed heights for easy optimization and scaling.
Specifications
| HiTrap Capto S ImpAct Prepacked Column | |
| 4 to 12 | |
| Ion Exchange Column |
| High-flow agarose | |
| 50μm |