missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Cytiva HisTrap™ FF Crude Prepacked Columns
Description
- Prepacked with Ni Sepharose™ 6 Fast Flow media
- Binding capacity approximately 40mg/mL medium with negligible Ni2+ leakage
- Maximum pressure: 3 bar
- Column material: polypropylene
- Compatible with wide variety of reducing agents, detergents, denaturants, and other additives
- 1 and 5mL columns
Media
- Matrix: 6% cross-linked agarose
- Average particle size: 90μm
- pH stability working range: 3 to 12
1mL columns
- Bed dimensions: 7 x 25mm
- Flow velocity: <4mL/min.
5mL columns
- Bed dimensions: 16 x 25mm
- Flow velocity: <20mL/min.
Specifications
Specifications
| Bed Volume | 1 mL |
| Diameter (English) Inner | 0.2 in. |
| Diameter (Metric) Inner | 7 mm |
| Description | HisTrap FF Crude columns are high binding capacity IMAC columns for his-tagged protein purification when scalability is needed. |
| Flow Rate | <4 mL/min. |
| Kit Contents | Sodium Phosphate-buffered Saline, pH 7.4 and 0.5X concentration of proprietary Y-PER reagent additives |
| Material | Polypropylene |
| pH Range | 3 to 12 |
| Support Type | 6% Cross-linked Agarose |
| Content And Storage | 4°C to 30°C |
| Show More |