missing translation for 'onlineSavingsMsg'
Learn More

HistoPrep™ Reagent Alcohol 80%

Catalog No. 11002216
Change view
Click to view available options
1 gal.
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No.
11002216 1 gal.
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Harmful if swallowed, in contact with skin, or if inhaled
Catalog No. 11002216 Supplier Statlab Supplier No. 11002216
Only null left
Harmful if swallowed, in contact with skin, or if inhaled

80% reagent alcohol.

  • 100% reagent alcohol with 20 parts Type 1 water.
  • Used as a dehydrating agent during tissue processing, gradually removing water from the tissue sample before it is embedded in paraffin for sectioning and staining

Chemical Identifiers

CAS 64-17-5 , 67-63-0 , 67-56-1

Specifications

Physical Form Aqueous Solution
Color Clear
Quantity 1 gal.
Grade Reagent

Made in the USA