missing translation for 'onlineSavingsMsg'
Learn More

HistoPrep™ Alcohol 95%

Catalog No. 11002223
Change view
Click to view available options
Quantity:
1 gal.
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
11002223 1 gal.
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Harmful if swallowed, in contact with skin, or if inhaled
Catalog No. 11002223 Supplier Statlab Supplier No. 11002223
Edge

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Harmful if swallowed, in contact with skin, or if inhaled

95% alcohol.

  • 100% alcohol with 5 parts deionized water.
  • Used to dehydrate tissue samples before embedding them in paraffin, effectively removing water from the tissue during the tissue processing stage

Chemical Identifiers

CAS 64-17-5 , 67-63-0 , 67-56-1

Specifications

Physical Form Aqueous Solution
Color Clear
Quantity 1 gal.
Grade Reagent

Made in the USA