missing translation for 'onlineSavingsMsg'
Learn More

HistoPrep™ Alcohol 70%

Catalog No. 11002219
Change view
Click to view available options
Quantity:
1 gal.
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
11002219 1 gal.
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Harmful if swallowed, in contact with skin, or if inhaled
Catalog No. 11002219 Supplier Statlab Supplier No. 11002219
Edge

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Harmful if swallowed, in contact with skin, or if inhaled

70% alcohol.

  • 100% reagent alcohol with 30 parts Type 1 water.
  • Used as a dehydrating agent during tissue processing, gradually removing water from the tissue sample before it is embedded in paraffin for sectioning and staining

Chemical Identifiers

CAS 64-17-5 , 67-63-0 , 67-56-1

Specifications

Physical Form Aqueous Solution
Color Clear
Quantity 1 gal.
Grade Reagent

Made in the USA