missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ HisPur™ Ni-NTA Resin

Catalog No. p-4531406
Change view
Click to view available options
Quantity:
10 mL
100 mL
500 mL
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PI88221 10 mL
PI88222 100 mL
PI88223 500 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. PI88221 Supplier Thermo Scientific™ Supplier No. 88221

Purify His-tagged recombinant fusion proteins using high-capacity nickel-NTA (nitrilotriacetic acid) metal-chelate beads, spin columns and plates.

Perform routine affinity purification of His-tagged fusion proteins with Thermo Scientific™ HisPur Ni-NTA Resin, a high-capacity, high-performance nickel-IMAC resin.

The specially prepared nickel-NTA support consists of beaded agarose derivatized with the nitrilotriacetic acid (NTA) chelation moiety and loaded with divalent nickel ions (Ni2+). The immobilized metal affinity chromatography (IMAC) resin provides exceptional binding capacity and performance for recombinant His-tagged protein purification.

HisPur Ni-NTA Resin is available in several package sizes of resin slurry, three sizes of centrifuge-ready columns, complete purification kits, two sizes of FPLC-ready chromatography cartridges, and 96-well filter plates for high through-put needs. With these options, high-yield His-tag protein purification is possible at nearly any scale.

Highlights:

  • High capacity – bind up to 60mg of 6xHis-tagged protein per milliliter of resin
  • Versatile – purify proteins using native or denaturing conditions
  • Compatible – use with Thermo Scientific Cell Lysis Reagents and a variety of buffer additives
  • Flexible – available in multiple formats including bulk resin, spin columns and chromatography cartridges
  • Cost-effective – reuse the same batch of resin at least five times
  • Easy to use – pre-formulated buffers available for kit formats

Available Formats:

  • Resin Slurries – crosslinked 6% beaded agarose; 10mL, 100mL, 500mL bottles
  • Spin Columns – 0.2mL (microcentrifuge), 1mL, 3mL columns
  • Purification Kits – complete kits in all three column sizes
  • Chromatography Cartridges – compatible with automated liquid chromatography systems and with syringe processing
  • Bulk and Custom – let us know your needs for bulk HisPur Ni-NTA Resin

Includes:

Kits contain HisPur Ni-NTA Columns (store at 4°C)

Specifications

Capacity (Metric) Up to 60 mg of 6xHis-Tagged Protein Binding per mL Resin
Description HisPur Ni-NTA Resin
Content And Storage Store at 4°C
Form Liquid Suspension
Product Line HisPur
Quantity 10 mL
Target His-Tagged Protein
Type Resin
Stationary Phase Divalent Nickel, Metal Chelate
Format Bottle
Column Type Affinity, Agarose Resin
Show More Show Less
Are there other sequences that can bind to nickel more tightly than 6xHis-tagged proteins and how can they be eluted.

Yes, 7xHis-tagged proteins, proteins naturally high in histidine, and other combinations of His and other amino acids will bind. To elute them, you have to increase the concentration of imidazole. Generally these peptides will not contaminate your fraction since they remain on the column. However after multiple uses of the same column, these peptides may reduce the binding capacity of the column.

Can the HisPur Ni-NTA Spin plates (Cat. No. 88230) be regenerated and reused, or are they for single-use only?

The resin can be re-used at least five times; this is also for the resin in the plates. Regeneration is possible using this procedure (https://assets.thermofisher.com/TFS-Assets%2FLSG%2Fmanuals%2FMAN0011700_HisPur_NiNTA_Resin_UG.pdf) Since the resin bed volume in each well is 50 µL, you can use 250 µL twice to give a total of 10 resin-bed volume.

How do I perform Ni-NTA bead regeneration with the HisPur Ni-NTA Resin (Cat. No. 88221)?

Please refer to page 2 (buffer preparation) and page 3 (procedure) of the manual (https://assets.thermofisher.com/TFS-Assets/LSG/manuals/MAN0011700_HisPur_NiNTA_Resin_UG.pdf) for information regarding bead regeneration.

Can ProBond or Ni-NTA beads be used for large-scale preparations?

ProBond and Ni-NTA beads can be used in FPLC columns. However, the beads can only withstand low pressure (~43.5 psi max).

What should the typical protein recovery be when using the Probond Purification System or Ni-NTA Purification system?

Both systems are qualified by purifying 2 mg of myoglobin protein on a column and performing a Bradford assay. Protein recovery must be 75% or higher.

What is the difference between HisPur Nickel Resin and HisPur Cobalt Resin?

Nickel has a higher affinity for histidines than does cobalt. It binds multiple histidines more tightly than cobalt, and requires more stringent conditions than is necessary for cobalt. The lower affinity of cobalt for multiple histidines typically results in less nonspecific binding of histidine-rich proteins that lack a his-tag compared to nickel resins.

HisPur Ni-NTA resin has a high capacity of up to 60 mg of 6xHis-tagged protein per milliliter. It is a versatile resin that can work under both native and denaturing conditions, and can also be used with a variety of lysis reagents and buffer additives.

HisPur Cobalt resin utilizes proprietary tetradentate chelating resin charged with cobalt. This system recovers highly purified protein with lower imidazole concentrations and has low metal leeching properties.


For Research Use Only. Not for use in diagnostic procedures.