Learn More
Thermo Scientific™ HisPur™ Cobalt Purification Kit, 0.2 mL
Description
Thermo Scientific™ HisPur Cobalt Resin is an immobilized metal chelate affinity (IMAC) support for specific, fast and gentle His-tagged purification.
Additional Information and Online Ordering
Highlights:
High purity – obtain more than 10 milligrams of pure His-tagged protein per milliliter of resin without optimizing imidazole washing conditions
Low metal leaching – no metal contamination in eluted histidine-tagged protein sample
Versatility – purify proteins under native or denaturing conditions; compatible with cell lysis reagents and a variety of buffer additives
Flexibility – available as bulk resin or predispensed columns, chromatography columns and 96-well filter plates
Cost effective – resin can be reused several times or discarded after single use
Superior – performs better than other commercially available IMAC Resins
Available Formats:
- Resin Slurries – crosslinked 6% beaded agarose; 10mL, 100mL, 500mL bottles
- Spin Columns – 0.2mL (microcentrifuge), 1mL, 3mL columns
- Purification Kits – complete kits in all three column sizes
- Chromatography Cartridges – compatible with automated liquid chromatography systems and with syringe processing
- 96-well Spin Plates – pre-dispensed 96-well filter plates, compatible with centrifugation and vacuum manifold systems for manual or automated purification. The plates enable fast, consistent well-to-well and plate-to-plate reproducibility for small-scale, high-throughput separations
- Bulk and Custom – let us know your needs for bulk HisPur Cobalt Resin
Includes:
Kits contain HisPur Cobalt Columns, equilibration/wash buffer and elution buffer
Specifications
Specifications
| Capacity (Metric) | 0.2 mL/Column |
| Description | HisPur Cobalt Spin Purification Kit, 0.2mL |
| Content And Storage | Sufficient For: Purifying ∼2 mg of His-tagged protein per column • HisPur Cobalt Spin Columns, 0.2 mL, 25 columns • Equilibration/Wash Buffer, 100 mL • Elution Buffer, 50 mL • Collection Tubes, 80 tubes Store at 4°C. |
| Product Line | HisPur |
| Quantity | 25 Column Kit |
| Target | His-Tagged Protein |
| Type | Protein Purification System |
| Stationary Phase | Metal Chelate, Divalent Cobalt |
| Sufficient For | 2 mg of His-Tagged Protein per column |
| Format | Kit, Microcentrifuge, Spin Column |
| Show More |
Frequently Asked Questions (FAQs)
While this is protein-dependent, many proteins have been successfully tested in downstream applications (reporter assays, immunoprecipitation and beta-Gal assays), including GST and 6xHis proteins. Otherwise, samples can be diluted or dialyzed to remove any interfering substances.
Nickel has a higher affinity for histidines than does cobalt. It binds multiple histidines more tightly than cobalt, and requires more stringent conditions than is necessary for cobalt. The lower affinity of cobalt for multiple histidines typically results in less nonspecific binding of histidine-rich proteins that lack a his-tag compared to nickel resins.
HisPur Ni-NTA resin has a high capacity of up to 60 mg of 6xHis-tagged protein per milliliter. It is a versatile resin that can work under both native and denaturing conditions, and can also be used with a variety of lysis reagents and buffer additives.
HisPur Cobalt resin utilizes proprietary tetradentate chelating resin charged with cobalt. This system recovers highly purified protein with lower imidazole concentrations and has low metal leeching properties.
The membrane used is regenerated cellulose with a low glycerol content (acts as a humectant).
For Research Use Only. Not for use in diagnostic procedures.