missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ HisPur™ Cobalt Chromatography Cartridges, 1 mL

Catalog No. PI90093
Change view
Click to view available options
Quantity:
2 cartridges x 5 mL
5 cartridges x 1 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PI90093 5 cartridges x 1 mL
PI90094 2 cartridges x 5 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. PI90093 Supplier Thermo Scientific™ Supplier No. 90093

Gently purify His-tagged recombinant fusion proteins using cobalt (Co2+) immobilized metal chelate affinity (IMAC) beads, spin columns and plates.

Thermo Scientific™ HisPur Cobalt Resin is an immobilized metal chelate affinity (IMAC) support for specific, fast and gentle His-tagged purification.

Additional Information and Online Ordering

HisPur Cobalt Resins, Columns and Kits use a tetradentate chelating resin charged with divalent cobalt (Co2+)to enable specific, fast and gentle purification of histidine-tagged fusion proteins from a variety of sample types. HisPur Cobalt Resin is available in several package sizes of resin slurry, three sizes of centrifuge-ready columns, complete purification kits, two sizes of FPLC-ready chromatography cartridges, and 96-well filter plates for high throughput needs. With these options, high-quality His-tag protein purification is possible at nearly any scale. HisPur Cobalt Resin is superior to traditional nickel-based immobilized metal affinity chromatography (IMAC) systems. Compared to Ni-IDA and tetradentate Ni2+chelate resins, HisPur Resin binds histidine-tagged proteins with higher specificity (less off-target binding) and releases them with gentler conditions (lower concentrations of imidazole). Use HisPur Columns and Cartridges to obtain high-purity His-tagged proteins with no metal contamination.

Highlights:

  • High purity – obtain more than 10 milligrams of pure His-tagged protein per milliliter of resin without optimizing imidazole washing conditions

  • Low metal leaching – no metal contamination in eluted histidine-tagged protein sample

  • Versatility – purify proteins under native or denaturing conditions; compatible with cell lysis reagents and a variety of buffer additives

  • Flexibility – available as bulk resin or predispensed columns, chromatography columns and 96-well filter plates

  • Cost effective – resin can be reused several times or discarded after single use

  • Superior – performs better than other commercially available IMAC Resins

Available Formats:

  • Resin Slurries – crosslinked 6% beaded agarose; 10mL, 100mL, 500mL bottles
  • Spin Columns – 0.2mL (microcentrifuge), 1mL, 3mL columns
  • Purification Kits – complete kits in all three column sizes
  • Chromatography Cartridges – compatible with automated liquid chromatography systems and with syringe processing
  • 96-well Spin Plates – pre-dispensed 96-well filter plates, compatible with centrifugation and vacuum manifold systems for manual or automated purification. The plates enable fast, consistent well-to-well and plate-to-plate reproducibility for small-scale, high-throughput separations
  • Bulk and Custom – let us know your needs for bulk HisPur Cobalt Resin

Includes:

Kits contain HisPur Cobalt Columns, equilibration/wash buffer and elution buffer

Specifications

Capacity (Metric) 1 mL
Content And Storage Store between 4°C and 8°C.
Form Liquid Suspension
Product Line HisPur
Quantity 5 cartridges x 1 mL
Target His-Tagged Protein
Type Chromatography Cartridge
Stationary Phase Metal Chelate, Divalent Cobalt
Column Type Affinity, Agarose Resin
What is the difference between HisPur Nickel Resin and HisPur Cobalt Resin?

Nickel has a higher affinity for histidines than does cobalt. It binds multiple histidines more tightly than cobalt, and requires more stringent conditions than is necessary for cobalt. The lower affinity of cobalt for multiple histidines typically results in less nonspecific binding of histidine-rich proteins that lack a his-tag compared to nickel resins.

HisPur Ni-NTA resin has a high capacity of up to 60 mg of 6xHis-tagged protein per milliliter. It is a versatile resin that can work under both native and denaturing conditions, and can also be used with a variety of lysis reagents and buffer additives.

HisPur Cobalt resin utilizes proprietary tetradentate chelating resin charged with cobalt. This system recovers highly purified protein with lower imidazole concentrations and has low metal leeching properties.

Are the rapid equilibrium dialysis (RED) Device units sterile or endotoxin-free?

No. The units do not undergo a sterilization procedure nor are they tested for endotoxin content.

For Research Use Only. Not for use in diagnostic procedures.