missing translation for 'onlineSavingsMsg'
Learn More

Cytiva HiScreen™ Sepharose™ Prepacked Fast Flow HIC Columns

Catalog No. 45002535
Change view
Click to view available options
Stationary Phase:
Butyl Sepharose
Butyl-S-Sepharose
Octyl Sepharose
Phenyl (low sub) Sepharose
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Stationary Phase
45002535 Butyl-S-Sepharose
45002534 Butyl Sepharose
45002538 Phenyl (low sub) Sepharose
45002536 Octyl Sepharose
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. 45002535 Supplier Cytiva Supplier No. 28926985

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Designed for method optimization and parameter screening

  • Ready-to-use columns for convenient process development optimization
  • Standard aliphatic/aromatic Hydrophobic Interaction Chromatography media for capture and intermediate purification
  • Bed height of 10cm designed to allow method optimization and parameter screening
  • Easily connected in series to achieve 20cm bed height
  • Small bed volume of 4.7mL for fast result and minimal sample/buffer consumption
  • Reproducible results, scalable to BioProcess columns packed with same chromatography media using same linear fluid velocity

  • Bed dimension: 7.7 × 100mm
  • Matrix: 4% cross-linked agarose/6% cross-linked agarose
  • pH stability working range (media): 3 to 13
  • Average particle size: 90μm
  • Material (column hardware and column tube): polypropylene
  • Column I.D.: 7.7mm
  • Pressure max.: 1.5 bar (22psi)
  • Flow velocity: <240/450cm/hr.

Specifications

Buffer 1mM HCl, 1M NaOH, 30% Isopropanol, 50% Ethylene Glycol, 70% Ethanol, 6M Guanidine Hydrochloride, 8M Urea
Product Type HiScreen Butyl-S-Sepharose 6 FF column
Bed Volume 4.7mL
Diameter (English) Inner 0.3 in.
Diameter (Metric) Inner 7.7mm
Quantity 1 x 4.7mL
Height (English) 3.93 in.
Height (Metric) 100mm
Flow Rate <450 cm/hr.
Material Polypropylene
Ligand Type Butyl-S
Max. Pressure 1.5 bar
pH Range 3 to 13
Particle Size 45 to 165μm
Support Type 6% Cross-linked Agarose
Content And Storage 4° to 30°C
Stationary Phase Butyl-S-Sepharose
Column Type Ion Exchange Column
For Use With (Application) Convenient process development optimization parameter screening
Show More Show Less