missing translation for 'onlineSavingsMsg'
Learn More

Cytiva HiScreen™ Q Sepharose™ and SP Sepharose™ High Performance IEX Columns

Designed for method optimization and parameter screening

$266.82 - $266.82

Specifications

Buffer 8M Urea, 6M Guanidine HCl, 70% Ethanol, 1M NaOH, 1M Acetic Acid, 30% Isopropanol, 30% Aceto nitrile, 2% SDS
Bed Volume 4.7mL
Diameter (English) Inner 0.3 in.
Diameter (Metric) Inner 7.7mm
Quantity 1 x 4.7mL
View More Specs

Products 2
Catalog Number Mfr. No. Product Type Ligand Type pH Range Price Quantity  
Catalog Number Mfr. No. Product Type Ligand Type pH Range Price Quantity  
45-002-540
View Documents
Cytiva
28950511
HiScreen Q Sepharose HP Column Quaternary Amine 2 to 12
Each for $266.82
Only null left
 
45-002-542
View Documents
Cytiva
28950515
HiScreen SP Sepharose HP Column Sulphopropyl 4 to 13
Each for $266.82
Only null left
 
Description

Description

  • Ready-to-use columns for convenient process development optimization and parameter screening
  • Highly cross-linked agarose matrix with an average bead size of 34μm
  • Bed height of 10cm designed to allow method optimization and parameter screening
  • Easily connected in series to achieve 20cm bed height
  • Small bed volume (4.7mL) gives fast result and minimal sample/buffer consumption
  • Reproducible results, scalable to BioProcess columns packed with the same chromatography media using the same linear fluid velocity

  • Bed dimension: 7.7 × 100mm
  • Matrix: 6% cross-linked agarose beads
  • Average particle size: 34μm
  • Max. flow velocity: <150cm/hr.
  • Material (column hardware and column tube): polypropylene
  • Column I.D.: 7.7mm
  • Flow velocity: <150cm/hr.

HiScreen Q HP

  • Ionic capacity: 0.14 to 0.20mmol Cl-/mL
  • Ligand: quaternary amine
  • Ion exchanger type: strong anion exchanger
HiScreen SP HP
  • Ionic capacity: 0.15 to 0.20mmol Cl-/mL
  • Ligand: sulphopropyl
  • Ion exchanger type: strong cation exchanger

Specifications

Specifications

8M Urea, 6M Guanidine HCl, 70% Ethanol, 1M NaOH, 1M Acetic Acid, 30% Isopropanol, 30% Aceto nitrile, 2% SDS
0.3 in.
1 x 4.7mL
100mm
Polypropylene
45 to 165μm
4° to 30°C
Optimization and parameter screening, ion exchange chromato graphy
4.7mL
7.7mm
3.93 in.
<150 cm/hr.
3 bar
6% Cross-linked Agarose
Ion Exchange Column
Videos
SDS
missing translation for 'documents'

missing translation for 'documents'