missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Cytiva His SpinTrap™ Spin Columns
Description
- Single-use spin columns prepacked with Ni Sepharose High Performance Medium that is precharged with nickel ions; negligible ion leakage
- One run in standard microcentrifuge takes approximatedly 10 min.
- Yields up to 750μg purified protein per column
- Allows direct purification of both clarified and unclarifed cell lysates
- Also useful for screening large numbers of small-scale bacterial lysates
- Kit option contains buffer, allowing fast and convenient purifications without time-consuming buffer preparation step
- Kit components (spin columns and buffer) can also be purchased separately
Medium Characteristics:
- Compatible with a wide range of reducing agents, detergents, denaturants and other additives
- Stable (long-term) in the pH range 3-12
- Average particle size: 34μm
Order Info
Kit (28-9321-71) contains 50 columns and one pack of His Buffer Kit (2 x 100mL phosphate 8X stock solution, pH 7.4, and 100mL highly pure 2M imidazole, pH 7.4)
Specifications
Specifications
| Kit Contents | 1 Pk. His SpinTrap (28-4013-53) and 1 Pk. His Buffer Kit (11-0034-00) |
| pH Range | 2 to 14 |
| Support Type | 6% Highly Cross-linked Agarose |
| Content And Storage | 4°C to 30°C |
| Quantity | 1 Piece |
| Type | Spin Column Kit |