Learn More
CoorsTek™ High-Form Porcelain Crucibles
Withstand temperatures up to 1150°C (2102°F), not to exceed 200°C/hour
$29.27 - $1,723.60
| Catalog Number | Mfr. No. | Capacity (Metric) | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Capacity (Metric) | Price | Quantity & Availability | |||||
|
07965H
|
CoorsTek™
CRS66110 |
100 mL |
Case of 24 Each for $1,723.60
|
|
|||||
|
07965C
|
CoorsTek™
CRS66104 |
10 mL |
Each for $29.27
Case of 72 Each for $2,107.44
|
|
|||||
|
07965G
|
CoorsTek™
CRS66109 |
80 mL |
Each for $63.43
Case of 24 Each for $1,522.32
|
|
|||||
|
079651
|
CoorsTek™
CRS66112 |
250 mL |
Each for $100.03
Case of 12 Each for $1,200.36
|
|
|||||
Description
- Glazed inside and out except for outside bottom surface and rim
- Covers sold separately
Specifications
| Porcelain |