missing translation for 'onlineSavingsMsg'
Learn More

Applied Biosystems™ High-Capacity RNA-to-cDNA™ Kit

Catalog No. 4388950
Encompass
Change view
Click to view available options
No. of Reactions:
50 Reactions
500 Reactions
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. No. of Reactions
4388950 500 Reactions
4387406 50 Reactions
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 4388950 Supplier Applied Biosystems™ Supplier No. 4388950

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Includes

1 x 500 uL enzyme mix (MuLV and RNase inhibitor protein) (20X), 5 x 1 mL RT Buffer Mix (2X) (includes dNTPs, random octamers, and oligo dT-16)

The High Capacity RNA-to-cDNA Kit is a streamlined reverse transcription kit designed for optimum performance with TaqMan Gene Expression Master Mix, Power SYBR Green PCR Master Mix, and other PCR enzymes.

The High Capacity RNA-to-cDNA Kit is a streamlined reverse transcription kit designed for optimum performance with TaqMan Gene Expression Master Mix, Power SYBR Green PCR Master Mix, and other PCR enzymes. The components of the two-tube kit work together to provide sensitive and specific reverse transcription across a broad range of template amounts.

Features of the High Capacity RNA-to-cDNA Kit include:
• Fast reaction time (typically 30–60 min)
• Convenient and simple workflow with few pipetting steps
• Reliable reverse transcription of both abundant and limited targets
• Optimized two-step protocol enables multiple PCR reactions from a single RT reaction

Streamline cDNA synthesis while maintaining detection sensitivity
Amplification of a dilution series of the RNA concentration reference standard demonstrates exceptional reverse transcription and PCR efficiency across 11 orders of magnitude. This wide dynamic range allows one set of reaction conditions to detect transcripts from highly active as well as weakly expressed genes. The two-tube formulation reduces reaction time and requires fewer pipetting steps than with the High Capacity cDNA Reverse Transcription Kit, while maintaining high performance.

Specifications

Content And Storage

• 20X Enzyme mix (1 x 500 μL)
• 2X RT Buffer Mix (5 x 1 mL)

Store at –15 to –25°C.
2X RT Buffer Mix can be stored at 2–8°C for up to 6 months.

Format Kit
Reaction Speed 30 to 60 min.
Technique Reverse Transcription
Optimal Reaction Temperature 37°C
Quantity 500 reactions
Reverse Transcriptase MultiScribe
Ribonuclease H Activity Yes
Shipping Condition Dry Ice
For Use With (Application) Real Time PCR (qPCR), RT-PCR
Final Product Type First-Strand cDNA
No. of Reactions 500 Reactions
Reaction Format Premixed Components
Reagent Type Reverse Transcription
Size (Final Product) Up to 7 kb
Starting Material RNA
Show More Show Less
Which reverse transcription kit can I use with the PowerUp SYBR Green Master Mix (Cat. No. A25741, A25742, A25743, A25776, A25777, A25778, A25779, A25780, A25919)?

We recommend using the High-Capacity RNA-to-cDNA Kit (Cat. No. 4387406, 4388950).

Which kit should I use for the reverse transcription step for TaqMan Pri-miRNA assays (Cat. No. 4427012)?

We recommend the following kits:

  • High-Capacity cDNA Reverse Transcription Kit (Cat. No. 4374966).
  • High-Capacity RNA-to-cDNA Kit (Cat. No. 4387406).

Is the 2X RT Buffer Mix within the High-Capacity RNA-to-cDNA Kit (Cat. N. 4387406, 4388950) available separately?

No, the 2X RT Buffer Mix is not available separately.

For Research Use Only. Not for use in diagnostic procedures.