missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MilliporeSigma Hi-Flow™ Plus Variety Sample Pack
Variety pack of lateral flow sheets.
$271.22 - $271.23
Specifications
| Filter Type | Membrane Filter |
|---|---|
| Format | Sheet |
| Material | Nitrocellulose |
| Sterility | Non-sterile |
| Flow Rate | 75 sec./4 cm, 90 sec./4 cm, 120 sec./4 cm, 135 sec./4 cm, 180 sec./4 cm |
| Catalog Number | Mfr. No. | Thickness (English) | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Thickness (English) | Price | Quantity & Availability | |||||
|
HFSAMPLE02
|
MilliporeSigma
HFSAMPLE02 |
2 mil |
Pack of 5 Each for $271.23
|
|
|||||
|
HFSAMPLE04
|
MilliporeSigma
HFSAMPLE04 |
4 mil |
Each for $271.22
|
|
|||||
Description
- Designed and optimised for use in rapid diagnostic lateral flow tests that require superior consistency and performance
- Lateral flow tests facilitate quick and convenient detection/quantification of analytes such as antibodies and antigens present in biological samples, including blood, saliva, and urine.
- Suitable for tests where speed is critical, the sample is plentiful, and sensitivity is not a concern
- Contains 10 sheets: 2 sheets each of Hi-Flow™ Plus 75, 90, 120, 135, and 180 membrane
- Dimensions: 30.5 x 25 cm
Specifications
| Membrane Filter | |
| Nitrocellulose | |
| 75 sec./4 cm, 90 sec./4 cm, 120 sec./4 cm, 135 sec./4 cm, 180 sec./4 cm | |
| Lateral Flow Membrane |
| Sheet | |
| Non-sterile | |
| 10 | |
| 30.5 x 25 cm |