missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MilliporeSigma Hi-Flow™ Plus Variety Sample Pack
Description
- Designed and optimised for use in rapid diagnostic lateral flow tests that require superior consistency and performance
- Lateral flow tests facilitate quick and convenient detection/quantification of analytes such as antibodies and antigens present in biological samples, including blood, saliva, and urine.
- Suitable for tests where speed is critical, the sample is plentiful, and sensitivity is not a concern
- Contains 10 sheets: 2 sheets each of Hi-Flow™ Plus 75, 90, 120, 135, and 180 membrane
- Dimensions: 30.5 x 25 cm
Spécifications
Spécifications
| Filter Type | Membrane Filter |
| Format | Sheet |
| Material | Nitrocellulose |
| Sterility | Non-sterile |
| Flow Rate | 75 sec./4 cm, 90 sec./4 cm, 120 sec./4 cm, 135 sec./4 cm, 180 sec./4 cm |
| Quantity | 10 |
| Thickness (English) | 4 mil |
| Type | Lateral Flow Membrane |
| Dimensions (L x W) | 30.5 x 25 cm |