missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Hexyl Mercaptan 96.0+%, TCI America™
$78.93 - $311.89
Chemical Identifiers
| CAS | 111-31-9 |
|---|---|
| Molecular Formula | C6H14S |
| Molecular Weight (g/mol) | 118.24 |
| MDL Number | MFCD00004909 |
| InChI Key | PMBXCGGQNSVESQ-UHFFFAOYSA-N |
| Synonym | 1-hexanethiol, hexyl mercaptan, n-hexyl mercaptan, hexanethiol, 1-hexylthiol, 1-mercaptohexane, hexylthiol, n-hexanethiol, n-hexylthiol, n-hexylmercaptan |
| PubChem CID | 8106 |
| IUPAC Name | hexane-1-thiol |
| SMILES | CCCCCCS |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|
H010325ML
|
TCI America
H010325ML |
25 mL |
Each for $78.93
|
|
|||||
|
H0103500ML
|
TCI America
H0103500ML |
500 mL |
Each for $311.89
|
|
|||||
Chemical Identifiers
| 111-31-9 | |
| 118.24 | |
| PMBXCGGQNSVESQ-UHFFFAOYSA-N | |
| 8106 | |
| CCCCCCS |
| C6H14S | |
| MFCD00004909 | |
| 1-hexanethiol, hexyl mercaptan, n-hexyl mercaptan, hexanethiol, 1-hexylthiol, 1-mercaptohexane, hexylthiol, n-hexanethiol, n-hexylthiol, n-hexylmercaptan | |
| hexane-1-thiol |
Specifications
| 111-31-9 | |
| Colorless | |
| C6H14S | |
| 25 mL | |
| 1-hexanethiol, hexyl mercaptan, n-hexyl mercaptan, hexanethiol, 1-hexylthiol, 1-mercaptohexane, hexylthiol, n-hexanethiol, n-hexylthiol, n-hexylmercaptan | |
| CCCCCCS | |
| 118.24 | |
| 118.24 | |
| Liquid |
| -81°C | |
| 150°C | |
| MFCD00004909 | |
| 1228 | |
| PMBXCGGQNSVESQ-UHFFFAOYSA-N | |
| hexane-1-thiol | |
| 8106 | |
| ≥96.0% (GC) | |
| Hexyl Mercaptan |
Safety and Handling
EINECSNumber : (2)-0464
RTECSNumber : MO4550000
TSCA : Yes