missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Hexyl Mercaptan 96.0+%, TCI America™
Supplier: TCI America H0103500ML
Specifications
| Hexyl Mercaptan | |
| -81°C | |
| 150°C | |
| MFCD00004909 | |
| 1228 | |
| PMBXCGGQNSVESQ-UHFFFAOYSA-N | |
| hexane-1-thiol | |
| 8106 | |
| ≥96.0% (GC) |
| 111-31-9 | |
| Colorless | |
| C6H14S | |
| 500 mL | |
| 1-hexanethiol, hexyl mercaptan, n-hexyl mercaptan, hexanethiol, 1-hexylthiol, 1-mercaptohexane, hexylthiol, n-hexanethiol, n-hexylthiol, n-hexylmercaptan | |
| CCCCCCS | |
| 118.24 | |
| 118.24 | |
| Liquid |
Chemical Identifiers
| 111-31-9 | |
| 118.24 | |
| PMBXCGGQNSVESQ-UHFFFAOYSA-N | |
| 8106 | |
| CCCCCCS |
| C6H14S | |
| MFCD00004909 | |
| 1-hexanethiol, hexyl mercaptan, n-hexyl mercaptan, hexanethiol, 1-hexylthiol, 1-mercaptohexane, hexylthiol, n-hexanethiol, n-hexylthiol, n-hexylmercaptan | |
| hexane-1-thiol |
Safety and Handling
EINECSNumber : (2)-0464
RTECSNumber : MO4550000
TSCA : Yes