missing translation for 'onlineSavingsMsg'
Learn More

Hexyl 4-Hydroxybenzoate 98.0+%, TCI America™

Catalog No. p-7130412
Change view
Click to view available options
Quantity:
25 g
100 g
500 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
H021325G 25 g
H0213100G 100 g
H0213500G 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. H021325G Supplier TCI America Supplier No. H021325G
Only null left

Chemical Identifiers

CAS 1083-27-8
Molecular Formula C13H18O3
Molecular Weight (g/mol) 222.284
MDL Number MFCD00016480
InChI Key ULULAZKOCFNOIM-UHFFFAOYSA-N
Synonym Hexylparaben, 4-Hydroxybenzoic Acid Hexyl Ester
PubChem CID 14127
IUPAC Name hexyl 4-hydroxybenzoate
SMILES CCCCCCOC(=O)C1=CC=C(C=C1)O

Specifications

Melting Point 53°C
Color White
Boiling Point 171°C
Quantity 25 g
Formula Weight 222.28
Percent Purity ≥98.0% (HPLC,T)
Physical Form Crystalline Powder
Chemical Name or Material Hexyl 4-Hydroxybenzoate
EINECSNumber (3)-1585
RTECSNumber DH2204000
TSCA No