accessibility menu, dialog, popup

UserName

H3COOOH

CAS RN 1083-27-8

IUPAC Name: hexyl 4-hydroxybenzoate
Synonyms: hexyl P-hydroxybenzoate
Molecular Weight (g/mol): 222.28
Molecular Formula: C13H18O3
InChi Key: ULULAZKOCFNOIM-UHFFFAOYSA-N
SMILES: CCCCCCOC(=O)C1=CC=C(O)C=C1
Percent Purity 
  • (3)
Physical Form 
  • (3)
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results