missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals Hexokinase, yeast

Catalog No. aaj63795jpl
Change view
Click to view available options
Quantity:
1 kU
10 kU
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAJ63795JPL 10 kU
AAJ637954I 1 kU
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAJ63795JPL Supplier Thermo Scientific Chemicals Supplier No. J63795JPL

Used to catalyzes the phosphorylation of D-hexose sugars at the C6 position utilizing, ATP as a phosphate source. Used to determine glucose, fructose, mannose and ATP.

Hexokinase is used to catalyzes the phosphorylation of D-hexose sugars at the C6 position utilizing, ATP as a phosphate source. Used to determine glucose, fructose, mannose and ATP.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Sensitivity Hygroscopic
Quantity 10 kU
Form Lyophilized

RUO – Research Use Only