missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Hexanoic Acid 98.0+%, TCI America™
Supplier: TCI America H010525ML
Specifications
| Hexanoic Acid | |
| -3°C | |
| 205°C | |
| MFCD00004421 | |
| 2829 | |
| FUZZWVXGSFPDMH-UHFFFAOYSA-N | |
| hexanoic acid | |
| 8892 | |
| 116.16 | |
| Liquid |
| 142-62-1 | |
| Yellow | |
| C6H12O2 | |
| 25 mL | |
| caproic acid, capronic acid, n-hexanoic acid, n-caproic acid, butylacetic acid, hexoic acid, pentylformic acid, 1-hexanoic acid, n-hexylic acid, n-hexoic acid | |
| CCCCCC(=O)O | |
| 116.16 | |
| CHEBI:30776 | |
| ≥98.0% (GC,T) |
Chemical Identifiers
| 142-62-1 | |
| 116.16 | |
| FUZZWVXGSFPDMH-UHFFFAOYSA-N | |
| 8892 | |
| hexanoic acid |
| C6H12O2 | |
| MFCD00004421 | |
| caproic acid, capronic acid, n-hexanoic acid, n-caproic acid, butylacetic acid, hexoic acid, pentylformic acid, 1-hexanoic acid, n-hexylic acid, n-hexoic acid | |
| CHEBI:30776 | |
| CCCCCC(=O)O |
Safety and Handling
EINECSNumber : (2)-0608
RTECSNumber : MO5250000
TSCA : Yes