missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Hexanoic Acid 98.0+%, TCI America™
$32.28 - $64.56
Chemical Identifiers
| CAS | 142-62-1 |
|---|---|
| Molecular Formula | C6H12O2 |
| Molecular Weight (g/mol) | 116.16 |
| MDL Number | MFCD00004421 |
| InChI Key | FUZZWVXGSFPDMH-UHFFFAOYSA-N |
| Synonym | caproic acid, capronic acid, n-hexanoic acid, n-caproic acid, butylacetic acid, hexoic acid, pentylformic acid, 1-hexanoic acid, n-hexylic acid, n-hexoic acid |
| PubChem CID | 8892 |
| ChEBI | CHEBI:30776 |
| IUPAC Name | hexanoic acid |
| SMILES | CCCCCC(=O)O |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
|
H010525ML
|
N/A | 25 mL |
Each for $32.28
|
|
|||||
|
H0105500ML
|
N/A | 500 mL |
Each for $64.56
|
|
|||||
Chemical Identifiers
| C6H12O2 | |
| MFCD00004421 | |
| caproic acid, capronic acid, n-hexanoic acid, n-caproic acid, butylacetic acid, hexoic acid, pentylformic acid, 1-hexanoic acid, n-hexylic acid, n-hexoic acid | |
| CHEBI:30776 | |
| CCCCCC(=O)O |
Specifications
| 142-62-1 | |
| Yellow | |
| C6H12O2 | |
| 2829 | |
| FUZZWVXGSFPDMH-UHFFFAOYSA-N | |
| hexanoic acid | |
| 8892 | |
| 116.16 | |
| Liquid |
| -3°C | |
| 205°C | |
| MFCD00004421 | |
| caproic acid, capronic acid, n-hexanoic acid, n-caproic acid, butylacetic acid, hexoic acid, pentylformic acid, 1-hexanoic acid, n-hexylic acid, n-hexoic acid | |
| CCCCCC(=O)O | |
| 116.16 | |
| CHEBI:30776 | |
| ≥98.0% (GC,T) | |
| Hexanoic Acid |
Safety and Handling
EINECSNumber : (2)-0608
RTECSNumber : MO5250000
TSCA : Yes