accessibility menu, dialog, popup

UserName

CAS RN 142-62-1

IUPAC Name: hexanoic acid
Synonyms: hexanoic acid hexanoic acid (natural) hexanoic acid, manganese (2+) salt rhodium (2+) salt hexanoic acid aliphatic carboxylic acid, c6:0 copper (2+) salt hexanoic acid hexanoic acid, rhodium (2+) salt pentiformic acid capronic acid hexanoic acid, nickel (2+) salt
Molecular Weight (g/mol): 116.16
Molecular Formula: C6H12O2
InChi Key: FUZZWVXGSFPDMH-UHFFFAOYSA-N
SMILES: CCCCCC(O)=O
Quantity 
  • (1)
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)
Boiling Point 
  • (3)
  • (4)
  • (2)
  • (1)
Percent Purity 
  • (3)
  • (2)
  • (1)
  • (4)
Physical Form 
  • (10)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (7)
  • (1)
  • (2)
  • (1)
  • (7)
  • (1)
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)
  • (3)
  • (4)
  • (2)
  • (1)
  • (3)
  • (2)
  • (1)
  • (4)
  • (10)