missing translation for 'onlineSavingsMsg'
Learn More

Hexane, Analytical Standard, ≥99.7% (GC), MilliporeSigma™ Supelco™

$301.93 - $1033.95

Specifications

CAS 110-54-3
Vapor Pressure 5.2 psi (37.7°C)
Vapor Density ˜3 (vs. air)
Density 0.659 g/mL (at 25°C)
Physical Form Neat
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
111019017
View Documents
Merck Emd Millipore
5275010ML
10 mL
Each for $301.93
Only null left
 
111018167
View Documents
Merck Emd Millipore
5275050ML
50 mL
Each for $1,033.95
Only null left
 
Description

Description

Specifications

Specifications

110-54-3
˜3 (vs. air)
Neat
MFCD00009520
69°C
86.18
Analytical Standard
5.2 psi (37.7°C)
0.659 g/mL (at 25°C)
CH3(CH2)4 CH3
-95°C
n-Hexane
≥99.7% (GC)
Videos
SDS
Documents

Documents