missing translation for 'onlineSavingsMsg'
Learn More

Hexane, Analytical Standard, ≥99.7% (GC), MilliporeSigma™ Supelco™

Catalog No. 111019017
Change view
Click to view available options
Quantity:
10 mL
50 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
111019017 10 mL
111018167 50 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 111019017 Supplier Merck Emd Millipore Supplier No. 5275010ML

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Specifications

CAS 110-54-3
Vapor Pressure 5.2 psi (37.7°C)
Vapor Density ˜3 (vs. air)
Density 0.659 g/mL (at 25°C)
Physical Form Neat
Linear Formula CH3(CH2)4 CH3
MDL Number MFCD00009520
Melting Point -95°C
Boiling Point 69°C
Synonym n-Hexane
Quantity 10 mL
Formula Weight 86.18
Percent Purity ≥99.7% (GC)
Grade Analytical Standard
Show More Show Less