missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Hexamethylbenzene 99.0+%, TCI America™
Supplier: TCI America H008725G
Specifications
| Hexamethylbenzene | |
| 165°C | |
| MFCD00008523 | |
| hexamethylbenzene, mellitene, benzene, hexamethyl, hexamethylbenzol, hexamethyl benzene, unii-j8sd5741v8, 1,2,3,4,5,6-hexamethyl-benzene, benzene, 1,2,3,4,5,6-hexamethyl, hexamethyl-benzene, hexamethylbenxene; | |
| CC1=C(C(=C(C(=C1C)C)C)C)C | |
| 162.276 | |
| CHEBI:39001 | |
| ≥99.0% (GC) |
| 87-85-4 | |
| C12H18 | |
| 25 g | |
| YUWFEBAXEOLKSG-UHFFFAOYSA-N | |
| 1,2,3,4,5,6-hexamethylbenzene | |
| 6908 | |
| 162.28 | |
| Crystalline Powder |
Chemical Identifiers
| 87-85-4 | |
| 162.276 | |
| YUWFEBAXEOLKSG-UHFFFAOYSA-N | |
| 6908 | |
| 1,2,3,4,5,6-hexamethylbenzene |
| C12H18 | |
| MFCD00008523 | |
| hexamethylbenzene, mellitene, benzene, hexamethyl, hexamethylbenzol, hexamethyl benzene, unii-j8sd5741v8, 1,2,3,4,5,6-hexamethyl-benzene, benzene, 1,2,3,4,5,6-hexamethyl, hexamethyl-benzene, hexamethylbenxene; | |
| CHEBI:39001 | |
| CC1=C(C(=C(C(=C1C)C)C)C)C |
Safety and Handling
RTECSNumber : DA3200000
TSCA : Yes