accessibility menu, dialog, popup

UserName

CH3CH3CH3CH3H3CH3C

CAS RN 87-85-4

IUPAC Name: hexamethylbenzene
Synonyms: hexamethylbenzene mellitene
Molecular Weight (g/mol): 162.28
Molecular Formula: C12H18
InChi Key: YUWFEBAXEOLKSG-UHFFFAOYSA-N
SMILES: CC1=C(C)C(C)=C(C)C(C)=C1C
Molecular Weight (g/mol) 
  • (2)
  • (6)
Percent Purity 
  • (3)
  • (2)
  • (1)
  • (2)
Quantity 
  • (1)
  • (3)
  • (3)
  • (1)
Boiling Point 
  • (2)
  • (3)
Physical Form 
  • (3)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (5)
  • (3)
  • (5)
  • (2)
  • (6)
  • (3)
  • (2)
  • (1)
  • (2)
  • (1)
  • (3)
  • (3)
  • (1)
  • (2)
  • (3)
  • (3)
  • (2)