missing translation for 'onlineSavingsMsg'
Learn More

Hexamethylbenzene 99.0+%, TCI America™

Catalog No. p-7130328
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
H00875G 5 g
H008725G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. H00875G Supplier TCI America Supplier No. H00875G
Only null left

Chemical Identifiers

CAS 87-85-4
Molecular Formula C12H18
Molecular Weight (g/mol) 162.276
MDL Number MFCD00008523
InChI Key YUWFEBAXEOLKSG-UHFFFAOYSA-N
Synonym hexamethylbenzene, mellitene, benzene, hexamethyl, hexamethylbenzol, hexamethyl benzene, unii-j8sd5741v8, 1,2,3,4,5,6-hexamethyl-benzene, benzene, 1,2,3,4,5,6-hexamethyl, hexamethyl-benzene, hexamethylbenxene;
PubChem CID 6908
ChEBI CHEBI:39001
IUPAC Name 1,2,3,4,5,6-hexamethylbenzene
SMILES CC1=C(C(=C(C(=C1C)C)C)C)C

Specifications

Melting Point 165°C
Quantity 5 g
Formula Weight 162.28
Percent Purity ≥99.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material Hexamethylbenzene
RTECSNumber DA3200000
TSCA Yes