Learn More
Hexafluoroacetone trihydrate, 98%
CAS: 34202-69-2 | C3H6F6O4 | 220.067 g/mol
Supplier: Thermo Scientific Chemicals L1077714
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
| Hexafluoroacetone trihydrate | |
| 18°C to 21°C | |
| 105°C to 106°C | |
| 1.319 | |
| 25 g | |
| 607236 | |
| SNZAEUWCEHDROX-UHFFFAOYSA-N | |
| 1,1,1,3,3,3-hexafluoropropan-2-one;trihydrate | |
| 36719 | |
| 98% |
| 34202-69-2 | |
| 1.579 | |
| C3H6F6O4 | |
| MFCD00149061 | |
| UN2552 | |
| hexafluoroacetone trihydrate, 1,1,1,3,3,3-hexafluoropropan-2-one trihydrate, 2-propanone, hexafluoro-, trihydrate, hfa trihydrate, acetone, hexafluoro-, trihydrate, 2-propanone, 1,1,1,3,3,3-hexafluoro-, trihydrate, perfluoroacetone trihydrate, hexafluoro-2-propanone trihydrate, hexafluoro-2-propanone trihydrate; perfluoroacetone trihydrate, hexafluoroacetonetrihydrate | |
| C(=O)(C(F)(F)F)C(F)(F)F.O.O.O | |
| 220.067 | |
| 220.07 (166.02 Anhydrous) |
Chemical Identifiers
| 34202-69-2 | |
| 220.067 | |
| SNZAEUWCEHDROX-UHFFFAOYSA-N | |
| 36719 | |
| C(=O)(C(F)(F)F)C(F)(F)F.O.O.O |
| C3H6F6O4 | |
| MFCD00149061 | |
| hexafluoroacetone trihydrate, 1,1,1,3,3,3-hexafluoropropan-2-one trihydrate, 2-propanone, hexafluoro-, trihydrate, hfa trihydrate, acetone, hexafluoro-, trihydrate, 2-propanone, 1,1,1,3,3,3-hexafluoro-, trihydrate, perfluoroacetone trihydrate, hexafluoro-2-propanone trihydrate, hexafluoro-2-propanone trihydrate; perfluoroacetone trihydrate, hexafluoroacetonetrihydrate | |
| 1,1,1,3,3,3-hexafluoropropan-2-one;trihydrate |
Safety and Handling
GHS H Statement
H301-H310-H330-H318-H361-H315-H335
Toxic if swallowed.
Fatal in contact with skin.
Fatal if inhaled.
Causes serious eye damage.
Suspected of damaging fertility or the unborn child.
Causes skin irritation.
May cause respiratory irritation.
P201-P202-P260-P262-P264b-P270-P271-P281-P284-P301+P310-P302+P350-P304+P340-P305+P351+P338-P308+P313-P310-P330-P332+P313-P361-P363-P501c
H301-H310+H330-H315-H318-H335-H361
DOTInformation : Transport Hazard Class: 6.1; Packing Group: II; Proper Shipping Name: HEXAFLUOROACETONE HYDRATE, LIQUID
EINECSNumber : 211-676-3
RTECSNumber : UC2700000
TSCA : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only