accessibility menu, dialog, popup

UserName

CAS RN 34202-69-2

H2OH2OH2OFFFOFFF

CAS RN 34202-69-2

IUPAC Name: hexafluoropropan-2-one trihydrate
Synonyms: hexafluoroacetone trihydrate
Molecular Weight (g/mol): 220.07
Molecular Formula: C3H6F6O4
InChi Key: SNZAEUWCEHDROX-UHFFFAOYSA-N
SMILES: O.O.O.FC(F)(F)C(=O)C(F)(F)F
Percent Purity 
  • (2)
  • (3)
Quantity 
  • (1)
  • (2)
  • (2)
Boiling Point 
  • (3)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (3)
  • (2)
  • (3)
  • (1)
  • (2)
  • (2)
  • (3)
  • (2)