missing translation for 'onlineSavingsMsg'
Learn More

Hexafluoroacetone trihydrate, 98%

Catalog No. p-7045450
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL1077706 5 g
AAL1077714 25 g
AAL1077722 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAL1077706 Supplier Thermo Scientific Chemicals Supplier No. L1077706
Only null left

CAS: 34202-69-2 | C3H6F6O4 | 220.067 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 34202-69-2
Molecular Formula C3H6F6O4
Molecular Weight (g/mol) 220.067
MDL Number MFCD00149061
InChI Key SNZAEUWCEHDROX-UHFFFAOYSA-N
Synonym hexafluoroacetone trihydrate, 1,1,1,3,3,3-hexafluoropropan-2-one trihydrate, 2-propanone, hexafluoro-, trihydrate, hfa trihydrate, acetone, hexafluoro-, trihydrate, 2-propanone, 1,1,1,3,3,3-hexafluoro-, trihydrate, perfluoroacetone trihydrate, hexafluoro-2-propanone trihydrate, hexafluoro-2-propanone trihydrate; perfluoroacetone trihydrate, hexafluoroacetonetrihydrate
PubChem CID 36719
IUPAC Name 1,1,1,3,3,3-hexafluoropropan-2-one;trihydrate
SMILES C(=O)(C(F)(F)F)C(F)(F)F.O.O.O

Specifications

Melting Point 18°C to 21°C
Density 1.579
Boiling Point 105°C to 106°C
Refractive Index 1.319
Quantity 5 g
UN Number UN2552
Beilstein 607236
Formula Weight 220.07 (166.02 Anhydrous)
Percent Purity 98%
Chemical Name or Material Hexafluoroacetone trihydrate
Hazard Category H301-H310+H330-H315-H318-H335-H361
Hazard Statement GHS H Statement
H301-H310-H330-H318-H361-H315-H335
Toxic if swallowed.
Fatal in contact with skin.
Fatal if inhaled.
Causes serious eye damage.
Suspected of damaging fertility or the unborn child.
Causes skin irritation.
May cause respiratory irritation.
Precautionary Statement P201-P202-P260-P262-P264b-P270-P271-P281-P284-P301+P310-P302+P350-P304+P340-P305+P351+P338-P308+P313-P310-P330-P332+P313-P361-P363-P501c
DOTInformation Transport Hazard Class: 6.1; Packing Group: II; Proper Shipping Name: HEXAFLUOROACETONE HYDRATE, LIQUID
EINECSNumber 211-676-3
RTECSNumber UC2700000
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only