missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Hexadecyltrimethylammonium Chloride 95.0+%, TCI America™
Supplier: TCI America H0082500G
Specifications
| Hexadecyltrimethylammonium Chloride | |
| 234°C | |
| 4.0 to 7.5 (50g/L H2O 25°C) | |
| MFCD00011773 | |
| 3077 | |
| WOWHHFRSBJGXCM-UHFFFAOYSA-M | |
| hexadecyltrimethylazanium chloride | |
| 8154 | |
| 320.00 | |
| Crystalline Powder |
| 112-02-7 | |
| White | |
| C19H42ClN | |
| 500 g | |
| hexadecyltrimethylammonium chloride, cetrimonium chloride, cetyltrimethylammonium chloride, n,n,n-trimethylhexadecan-1-aminium chloride, dehyquart a, cetyl trimethyl ammonium chloride, aliquat 6, trimethylcetylammonium chloride, arquad 16-50, n-hexadecyltrimethylammonium chloride | |
| [Cl-].CCCCCCCCCCCCCCCC[N+](C)(C)C | |
| 320.00 | |
| CHEBI:53581 | |
| ≥95.0% (T) |
Chemical Identifiers
| 112-02-7 | |
| 320.00 | |
| WOWHHFRSBJGXCM-UHFFFAOYSA-M | |
| 8154 | |
| hexadecyltrimethylazanium chloride |
| C19H42ClN | |
| MFCD00011773 | |
| hexadecyltrimethylammonium chloride, cetrimonium chloride, cetyltrimethylammonium chloride, n,n,n-trimethylhexadecan-1-aminium chloride, dehyquart a, cetyl trimethyl ammonium chloride, aliquat 6, trimethylcetylammonium chloride, arquad 16-50, n-hexadecyltrimethylammonium chloride | |
| CHEBI:53581 | |
| [Cl-].CCCCCCCCCCCCCCCC[N+](C)(C)C |
Safety and Handling
EINECSNumber : (2)-0184&(9)-1971
RTECSNumber : ML9145000
TSCA : Yes